Titanium(II) hydride, fine powder, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.12384.0100 1.12384.1000 1.12384.0100 1.12384.1000
EM1.12384.0100CS 4378.18 CAD
EM1.12384.0100 EM1.12384.1000 CA1.12384.0100 CA1.12384.1000
Titanium(II) hydride, fine powder, Sigma-Aldrich®
Titanium (II) hydride
Formula: H₂Ti
Density: 3.91 g/cm³ (20 °C)
MDL Number: MFCD00151301
CAS Number: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,
Merck Index: 14,09473

Order Now

Specification Test Results

Melting Point 400 °C
Molar Mass 49.88 g/mol
Bulk Density 1400 kg/m3
Flash Point 400 °C
Density 3.76 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 400 °C
Hazard Class 4.1
Hazard Class Description Flammable Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR