1-Octanesulfonic acid sodium salt for surfactant analysis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.12292.0005 1.12292.0025 1.12292.0005 1.12292.0025
EM1.12292.0005CS 8810.08 CAD
EM1.12292.0005 EM1.12292.0025 CA1.12292.0005 CA1.12292.0025
1-Octanesulfonic acid sodium salt for surfactant analysis, Sigma-Aldrich®
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 216.27 g/mol
Bulk Density 345 kg/m3
pH 5.5 - 7.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR