Lanthanum(III) chloride heptahydrate ≥98% for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ100
1.12219.0100
1.12219.0250
CA1.12219.0100CS
1222
CAD
CA1.12219.0100
CA1.12219.0250
Lanthanum(III) chloride heptahydrate ≥98% for analysis, Supelco®
Lanthanum (III) chloride heptahydrate
1.12219.0100 replaces and is the same as LX0042-3. Part # Change only, Same Pack Size, Same Specifications.
Formula:
LaCl₃·7H₂O MW: 371.37 g/mol Boiling Pt: ∼860 °C (1013 hPa) Melting Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149756 CAS Number: 10025-84-0 EINECS: 233-237-5 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 91 °C Elimination of water of crystallisation |
Molar Mass | 371.37 g/mol |
Bulk Density | 900 kg/m3 |
pH | 5 (100 g/l, H2O, 25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...