1-Dodecansulfonic acid sodium salt for tenside tests, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.12146.0005 1.12146.0005 1.12146.8500
EM1.12146.0005CS 18555.52 CAD
EM1.12146.0005 CA1.12146.0005 CA1.12146.8500
1-Dodecansulfonic acid sodium salt for tenside tests, Sigma-Aldrich®
1-Dodecansulphonic acid sodium salt
Formula: C₁₂H₂₅NaO₃S
Melting Pt: 300 °C
Density: 1.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0
EINECS: 219-200-6

Order Now

Specification Test Results

Assay (alkalimetric, calculated on dried substance) ≥ 99.0 %
Identity (IR-spectrum) conforms
Loss on drying (120 °C; 4h; Vacuum) ≤ 2.0 %
pH (100\u2007g/l, water) 5.5 - 7.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR