di-Lithium tetraborate 99,0 Acidimetry analysis_for, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Lithium tetraborate, Dilithium tetraborate
MQ100
1.10783.1000
1.10783.5000
1.10783.9025
CA1.10783.1000EA
991
CAD
CA1.10783.1000
CA1.10783.5000
CA1.10783.9025
di-Lithium tetraborate 99,0 Acidimetry analysis_for, Supelco®
di-Lithium tetraborate
EMD # 1.10783.1000 replaces and is identical to EMD # 10783-3.
Formula:
Li₂B₄O₇ MW: 169.12 g/mol Melting Pt: 918 Density: 2.35 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011083 CAS Number: 12007-60-2 EINECS: 234-514-3 Merck Index: 14,05524 |
Specification Test Results
Assay (acidimetric) | ≥ 98.0 % |
Chloride (Cl) | ≤ 0.005 % |
Phosphate (PO₄) | ≤ 0.002 % |
Sulfate (SO₄) | ≤ 0.005 % |
Heavy metals (as Pb) | ≤ 0.005 % |
Ca (Calcium) | ≤ 0.005 % |
Fe (Iron) | ≤ 0.001 % |
K (Potassium) | ≤ 0.005 % |
Mg (Magnesium) | ≤ 0.001 % |
Na (Sodium) | ≤ 0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...