Sodium thiosulfate 0.01 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma

Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfiteAntichlorSodiumthiosulphate

1.09909.0001 1.09909.0001
CA1.09909.0001CS 6531.85 CAD
EM1.09909.0001 CA1.09909.0001
Sodium thiosulfate 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.02 g/cm3 (20 °C)
pH 9 - 10 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR