Ferroin (1,10-phenanthrolineferrous sulfate complex) 1/40 mol/L in aqueous solution redox indicator, Supelco®

Supplier: MilliporeSigma

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate, Tris(1,10-phenanthroline)iron (II) sulfate, Tris(1,10-phenanthroline-N1,N10)iron sulfate

1.09193.0100 1.09193.0500 1.09193.0100 1.09193.0500
EM1.09193.0100CS 1777.5 CAD
EM1.09193.0100 EM1.09193.0500 CA1.09193.0100 CA1.09193.0500
Ferroin (1,10-phenanthrolineferrous sulfate complex) 1/40 mol/L in aqueous solution redox indicator, Supelco®
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)
Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00036428
CAS Number: 14634-91-4
EINECS: 238-676-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 2.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR