Fluorescent indicator F₂₅₄, Millipore®

Supplier: MilliporeSigma

1.09182.0050 1.09182.0050
CA1.09182.0050CS 40110.96 CAD
CA1.09182.0050 EM1.09182.0050
Fluorescent indicator F₂₅₄, Millipore®
Fluorescent indicator F₂₅₄

EMD # 1.09182.0050 replaces and is identical to EMD # 9182-1.

Application: Fluorescent indicator F₂₅₄. CAS 68611-47-2, pH 7.0 (H₂O) suspension.
Physicochemical Information
Density: 4.0 g/cm³ (20 °C)
Melting Point: 1500 °C
pH value: 7.0 (H₂O) suspension
Bulk density: 1450 kg/m³
Solubility: <0.1 g/L

CAS Number: 68611-47-2

Order Now

Specification Test Results

Identity Conforms
Fe (iron) ≤0,01%
Water ≤0,3%
Application test for DC Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR