Iodine monochloride solution 0.1 mol/l acc. Wijs, Titripur® for determination of iodine value, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Iodochloride

M-Clarity™ quality level = MQ100

1.09163.1000 1.09163.2500
CA1.09163.1000CS 907 CAD
CA1.09163.1000 CA1.09163.2500
Iodine monochloride solution 0.1 mol/l acc. Wijs, Titripur® for determination of iodine value, Supelco®
Iodine monochloride
Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00011354
CAS Number: 7790-99-0
UN: 2789
ADR: 8 (3) II
Merck Index: 13,05039

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C. Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR