Zinc chloride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

1.08816.0250 1.08816.1000
CA1.08816.0250EA 242 CAD
CA1.08816.0250 CA1.08816.1000
Zinc chloride, EMSURE® for analysis, Supelco®
Zinc chloride

1.08816.0250 is offered as an alternative to ZX0065-1. Different Pack Size, Same Specifications.
1.08816.1000 is offered as an alternative to ZX0065-5. Different Pack Size, Same Specifications.

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185

Order Now

Specification Test Results

Solubility 4320 g/l (25 °C)
Melting Point 283 °C
Molar Mass 136.30 g/mol
Bulk Density 1400 - 1800 kg/m3
Boiling Point 732 °C (1013 hPa)
Vapor Pressure 1.33 hPa (428 °C)
Density 2.91 g/cm3 (25 °C)
pH 5 (100 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR