Zinc acetate dihydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

1.08802.0250 1.08802.1000
CA1.08802.0250EA 148 CAD
CA1.08802.0250 CA1.08802.1000
Zinc acetate dihydrate, EMSURE® for analysis, Supelco®
Zinc acetate dihydrate

1.08802.0250 replaces and is the same as ZX0050-2. Part # Change only, Same Pack Size, Same Specifications.

Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Order Now

Specification Test Results

Solubility 430 g/l (20 °C)
Melting Point 237 °C
Molar Mass 219.49 g/mol
Bulk Density 900 kg/m3
Density 1.74 g/cm3 (20 °C)
pH 6 - 7 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR