Xylenol orange tetrasodium salt indicator for metal titration, Supelco®

Supplier: MilliporeSigma

1.08677.0001 1.08677.0005 1.08677.0001 1.08677.0005
EM1.08677.0001CS 18628.2 CAD
EM1.08677.0001 EM1.08677.0005 CA1.08677.0001 CA1.08677.0005
Xylenol orange tetrasodium salt indicator for metal titration, Supelco®
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
Melting Pt: ∼210 °C
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now

Specification Test Results

Solubility 510 g/l (20 °C)
Molar Mass 760.59 g/mol
Bulk Density 250 kg/m3
pH 8.1 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR