Titriplex® II (EDTA (Ethylenediamine tetraacetic acid)) 99.4-100.6% (by complexometric analysis) ACS, Reag. Ph. Eur., Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

1.08417.0100 1.08417.0250 1.08417.1000
CA1.08417.0100CS 5615 CAD
CA1.08417.0100 CA1.08417.0250 CA1.08417.1000 CA1.08417.5000
Titriplex® II (EDTA (Ethylenediamine tetraacetic acid)) 99.4-100.6% (by complexometric analysis) ACS, Reag. Ph. Eur., Supelco®
EDTA (Ethylenediamine tetraacetic acid)
Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614
Melting Pt: 220
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (complexometric) 99.4 - 100.6 %
Identity (IR-spectrum) conforms
Appearance White to almost white, fine crystalline to crystalline powder.
in ammonia insoluble components ≤ 0.005 %
Heavy metals (as Pb) ≤ 10 ppm
Ca (Calcium) ≤ 10 ppm
Fe (Iron) ≤ 10 ppm
Mg (Magnesium) ≤ 5 ppm
Nitrilotriacetic acid ≤ 0.1 %
Sulfated ash ≤ 0.1 %
Water (according to Karl Fischer) ≤ 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR