Thymol blue, Supelco®
Supplier: MilliporeSigma
1.08176.0005
1.08176.0025
1.08176.0005
1.08176.0025
EM1.08176.0005CS
7825.74
CAD
EM1.08176.0005
EM1.08176.0025
CA1.08176.0005
CA1.08176.0025
Thymol blue, Supelco®
Thymol blue
Formula:
C₂₇H₃₀O₅S Melting Pt: 221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005869 CAS Number: 76-61-9 EINECS: 200-973-3 Merck Index: 12,09541 |
Specification Test Results
Solubility | 0.11 g/l (25 °C) |
Melting Point | 221 °C (decomposition) |
Molar Mass | 466.59 g/mol |
Bulk Density | 350 kg/m3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...