Thymolphthalein indicator, Supelco®

Supplier: MilliporeSigma

1.08175.0005 1.08175.0025 1.08175.0100 1.08175.0005 1.08175.0025 1.08175.0100
EM1.08175.0005CS 7571.36 CAD
EM1.08175.0005 EM1.08175.0025 EM1.08175.0100 CA1.08175.0005 CA1.08175.0025 CA1.08175.0100
Thymolphthalein indicator, Supelco®
Thymolphthaleine
Formula: C₂₈H₃₀O₄
Melting Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
EINECS: 204-729-7
Merck Index: 12,09542

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 253 °C
Molar Mass 430.54 g/mol
Bulk Density 400 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR