Bromophenol blue indicator, Supelco®

Supplier: MilliporeSigma

1.08122.0005 1.08122.0025 1.08122.0005 1.08122.0025
EM1.08122.0005CS 35924.46 CAD
EM1.08122.0005 EM1.08122.0025 CA1.08122.0005 CA1.08122.0025
Bromophenol blue indicator, Supelco®
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
Melting Pt: 273 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 273 °C (decomposition)
Molar Mass 669.96 g/mol
Bulk Density 730 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR