Strontium chloride hexahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.07865.0250 1.07865.1000
CA1.07865.0250EA 247 CAD
CA1.07865.0250 CA1.07865.1000
Strontium chloride hexahydrate, EMSURE® for analysis, Supelco®
Strontium chloride hexahydrate
Formula: SrCl₂·6H₂O
MW: 266.62 g/mol
Boiling Pt: 100
Melting Pt: 61
Density: 1.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149865
CAS Number: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Order Now

Specification Test Results

Solubility 2058 g/l (40 °C)
Melting Point 61 °C
Molar Mass 266.62 g/mol
Bulk Density 1050 kg/m3
Boiling Point 100 °C
Density 1.95 g/cm3 (20 °C)
pH 5 - 7 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR