Tin(II) chloride dihydrate, EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Stannous chloride dihydrate

1.07815.0100 1.07815.0250 1.07815.1000
CA1.07815.0100EA 259 CAD
CA1.07815.0100 CA1.07815.0250 CA1.07815.1000
Tin(II) chloride dihydrate, EMSURE® analysis_for, Supelco®
Tin (II) chloride dihydrate

1.07815.0250 is offered as an alternative to SX0885-1. Different Pack Size, Same Specifications.
1.07815.1000 is offered as an alternative to SX0885-5. Different Pack Size, Same Specifications.

Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623
Melting Pt: 246
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 II
Merck Index: 13,08861

Order Now

Specification Test Results

Solubility 1187 g/l (20 °C)
Melting Point 246 °C (anhydrous substance)
Molar Mass 225.63 g/mol
Bulk Density 1250 kg/m3
Boiling Point 623 °C (1013 hPa) (anhydrous substance)
Density 2.71 g/cm3 (20 °C)
pH 1 - 2 (100 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR