Silica Gel 60 H for Thin-Layer Chromatography, Merck
Supplier: MilliporeSigma
1.07736.1000 1.07736.2500 1.07736.9025 1.07736.1000 1.07736.2500 1.07736.9025
EM1.07736.1000EA 337.61 CAD
EM1.07736.1000 EM1.07736.2500 EM1.07736.9025 CA1.07736.1000 CA1.07736.2500 CA1.07736.9025
Silica Gel 60 H for Thin-Layer Chromatography, Merck
Chromatography Sorbents TLC Sorbents
Standardized sorbents ensure reliable results every time
EMD # 1.07736.9025 replaces and is identical to EMD # 7736-9.
EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).