Silica Gel 60 GF₂₅₄ for Thin-Layer Chromatography, Merck
Supplier: MilliporeSigma
1.07730.5000 1.07730.9025 1.07730.1000 1.07730.1000 1.07730.0500 1.07730.0500 1.07730.5000 1.07730.9025
CA1.07730.1000CS 2489.97 CAD
CA1.07730.1000 EM1.07730.0500 CA1.07730.0500 CA1.07730.9025 EM1.07730.5000 EM1.07730.9025 CA1.07730.5000 EM1.07730.1000
Silica Gel 60 GF₂₅₄ for Thin-Layer Chromatography, Merck
Chromatography Sorbents TLC Sorbents
Standardized sorbents ensure reliable results every time
EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).