Sodium sulfite, anhydrous 97,0-100,5 Iodometry, EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma

1.06657.0500 1.06657.1000 1.06657.5000 1.06657.9050
CA1.06657.0500EA 153 CAD
CA1.06657.0500 CA1.06657.1000 CA1.06657.5000 CA1.06657.9050
Sodium sulfite, anhydrous 97,0-100,5 Iodometry, EMSURE® analysis_for, Supelco®
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Solubility 220 g/l (20 °C)
Melting Point 500 °C (decomposition)
Molar Mass 126.04 g/mol
Bulk Density 1480 kg/m3
Density 2.63 g/cm3 (20 °C)
pH 8.8 - 10 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR