Sodium sulfate, anhydrous ≥99.0% (by acidimetry), coarse granules, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
1.06637.0500
1.06637.1000
1.06637.9025
CA1.06637.0500EA
96
CAD
CA1.06637.0500
CA1.06637.1000
CA1.06637.9025
Sodium sulfate, anhydrous ≥99.0% (by acidimetry), coarse granules, EMSURE® for analysis, Supelco®
Sodium sulphate
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 884 °C (lit.) Density: 2.68 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 12,08829 |
|
Specification Test Results
Solubility | 44.55 g/l (20 °C) |
Melting Point | 888 °C |
Molar Mass | 142.04 g/mol |
Bulk Density | 1400 - 1600 kg/m3 |
Density | 2.70 g/cm3 (20 °C) |
pH | 4 - 7 (200 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...