Sodium salicylate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

1.06601.0250 1.06601.1000
CA1.06601.0250EA 200 CAD
CA1.06601.0250 CA1.06601.1000
Sodium salicylate, EMSURE® for analysis, Supelco®
Sodium salicylate

In North America 1.06601.0250 replaces but with a superior specification and smaller pack size to SX0745-1
In North America 1.06601.1000 replaces but with a superior specification and smaller pack size to SX0745-3

Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: 200 °C
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point 200 °C
Molar Mass 160.10 g/mol
Bulk Density 320 kg/m3
Density 0.32 g/cm3 (20 °C)
pH 6.5 (100 g/l, H2O, 20 °C)
Ignition Point 250 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR