Sodium periodate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium metaperiodate

1.06597.0050 1.06597.0250 1.06597.1000
CA1.06597.0050EA 326 CAD
CA1.06597.0050 CA1.06597.0250 CA1.06597.1000
Sodium periodate, EMSURE® for analysis, Supelco®
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300
Melting Pt: 175
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1 (8) I
Merck Index: 13,08714

Order Now

Specification Test Results

Solubility 91 g/l (20 °C)
Melting Point 175 °C decomposes
Molar Mass 213.89 g/mol
Bulk Density 2000 - 2400 kg/m3
Boiling Point 300 °C
Density 3.87 g/cm3 (20 °C)
pH 5.2 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR