Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
1.06580.9050
1.06580.0500
1.06580.1000
1.06580.5000
1.06580.9025
CA1.06580.9050EA
7083
CAD
CA1.06580.9050
CA1.06580.0500
CA1.06580.1000
CA1.06580.5000
CA1.06580.9025
Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis, Supelco®
di-Sodium hydrogen phosphate dihydrate
1.06580.5000 replaces and is the same as SX0713-5. Part # Change only, Same Pack Size, Same Specifications.
Formula:
Na₂HPO₄·2H₂O Melting Pt: 92.5 °C (–H₂O of crystall.) Density: 2.1 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149182 CAS Number: 10028-24-7 EINECS: 231-448-7 UN: 3077 |
Specification Test Results
Assay (acidimetric) | ≥ 99.5 % |
Identity | passes test |
pH-value (5 %; water) | 9.0 - 9.2 |
Chloride (Cl) | ≤ 0.001 % |
Sulfate (SO₄) | ≤ 0.005 % |
Total nitrogen (N) | ≤ 0.001 % |
Heavy metals (as Pb) | ≤ 0.001 % |
Cu (Copper) | ≤ 0.0003 % |
Fe (Iron) | ≤ 0.001 % |
K (Potassium) | ≤ 0.005 % |
Pb (Lead) | ≤ 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...