Sodium phosphate dibasic, anhydrous, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
Synonyms:
di-Sodium hydrogen phosphate anhydrous
, Sodium phosphate dibasic
, sec-Sodium phosphate
, Disodium phosphate
, di-Sodium hydrogen orthophosphate
, Sodium hydrogen phosphate
1.06566.0500
1.06566.0100
1.06566.0500
1.06566.0100
CA1.06566.0100EA
578.82
CAD
EM1.06566.0100
EM1.06566.0500
CA1.06566.0100
CA1.06566.0500
Sodium phosphate dibasic, anhydrous, Suprapur® for trace metal analysis, Supelco®
di-Sodium hydrogen phosphate
Formula:
Na₂HPO₄ MW: 141.96 g/mol Melting Pt: 250 °C Density: 1.53 g/cm³ (20 °C) |
MDL Number:
MFCD00003496 CAS Number: 7558-79-4 Merck Index: 12,08805 |
Specification Test Results
Solubility | 77 g/l (20 °C) |
Melting Point | 250 °C (decomposition) |
Molar Mass | 141.96 g/mol |
Bulk Density | 880 kg/m3 |
pH | 8.7 - 9.3 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...