Sodium perchlorate monohydrate 99.0-102.0% (by alkalimetry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.06564.0100
1.06564.0500
1.06564.9025
CA1.06564.0100EA
119
CAD
CA1.06564.0100
CA1.06564.0500
CA1.06564.9025
Sodium perchlorate monohydrate 99.0-102.0% (by alkalimetry), EMSURE® for analysis, Supelco®
Sodium perchlorate monohydrate
1.06564.0500 replaces and is the same as SX0693-2. Part # Change only, Same Pack Size, Same Specifications.
1.06564.0500 is offered as an alternative to SX0693-1. Different Pack Size, Same Specifications.
Formula:
NaClO₄·H₂O Melting Pt: 130 °C Density: 2.02 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149164 CAS Number: 7791-07-3 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 12,08798 |
Specification Test Results
Assay (alkalimetric) | 99.0 - 102.0 % |
pH-value (5 %; water) | 4.5 - 7.0 |
Chloride (Cl) | ≤ 0.002 % |
Chloride, Chlorate (as Cl) | ≤ 0.002 % |
Sulfate (SO₄) | ≤ 0.002 % |
Total nitrogen (N) | ≤ 0.0005 % |
Ca (Calcium) | ≤ 0.002 % |
Fe (Iron) | ≤ 0.0003 % |
K (Potassium) | ≤ 0.005 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...