Sodium peroxide, granular for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06563.0100 1.06563.0500 1.06563.1000
CA1.06563.0100EA 184 CAD
CA1.06563.0100 CA1.06563.0500 CA1.06563.1000
Sodium peroxide, granular for analysis, Supelco®
Sodium peroxide

1.06563.1000 replaces and is the same as SX0700-6. Part # Change only, Same Pack Size, Same Specifications.

Formula: Na₂O₂
Melting Pt: 660 °C
Density: 2.805 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 13,08728

Order Now

Specification Test Results

Solubility 100 g/l (20 °C) (decomposition)
Melting Point 660 °C
Molar Mass 77.98 g/mol
Bulk Density 1138 kg/m3
Density 2.8 g/cm3 (20 °C)
pH 12.8 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR