Disodium oxalate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

1.06557.0250 1.06557.1000
CA1.06557.0250EA 236 CAD
CA1.06557.0250 CA1.06557.1000
Disodium oxalate, EMSURE® for analysis, Supelco®
di-Sodium oxalate
Formula: Na₂C₂O₄
MW: 134 g/mol
Melting Pt: 250…270 °C
Density: 2.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Order Now

Specification Test Results

Solubility 37 g/l (20 °C)
Melting Point 250 - 270 °C (decomposition)
Molar Mass 134 g/mol
Bulk Density 600 kg/m3
Density 2.27 g/cm3 (20 °C)
pH 8 (30 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR