Sodium metabisulfite 98.0-100.5% (by iodometry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06528.0100 1.06528.0500 1.06528.1000 1.06528.5000 1.06528.9050
CA1.06528.0100EA 63 CAD
CA1.06528.0100 CA1.06528.0500 CA1.06528.1000 CA1.06528.5000 CA1.06528.9050
Sodium metabisulfite 98.0-100.5% (by iodometry), EMSURE® for analysis, Supelco®
Sodium metabisulphite
Formula: Na₂S₂O₅
Melting Pt: 150 °C (dec.)
Density: 2.36 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
UN: 3077
REACH: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 650 g/l (20 °C)
Melting Point 150 °C (decomposition)
Molar Mass 190.11 g/mol
Bulk Density 1000 - 1200 kg/m3
Density 2.36 g/cm3 (20 °C)
pH 4.0 - 4.8 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR