Sodium molybdate(VI) dihydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

1.06521.0100 1.06521.0250 1.06521.1000
CA1.06521.0100EA 234 CAD
CA1.06521.0100 CA1.06521.0250 CA1.06521.1000
Sodium molybdate(VI) dihydrate, EMSURE® for analysis, Supelco®
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Melting Pt: 687
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Solubility 840 g/l (20 °C)
Melting Point 687 °C Elimination of water of crystallisation
Molar Mass 241.95 g/mol
Bulk Density 1000 - 1400 kg/m3
Density 3.28 g/cm3 (20 °C) (for the dihydrate)
pH 7.9 - 10.3 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR