Sodium hydroxide ≥99% (NaOH basis by acidimetry), granulated, Supelco®

Supplier: MilliporeSigma
Danger

1.06467.9010 1.06467.9050
CA1.06467.9010EA 728 CAD
CA1.06467.9010 CA1.06467.9050
Sodium hydroxide ≥99% (NaOH basis by acidimetry), granulated, Supelco®
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 318 °C (lit.)
Density: 2.13 g/mL (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Solubility 1090 g/l (20 °C)
Melting Point 323 °C
Molar Mass 40.00 g/mol
Boiling Point 1390 °C (1013 hPa)
Vapor Pressure (20 °C)
Density 2.13 g/cm3 (20 °C)
pH 14 (50 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR