Sodium hydroxide monohydrate, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.06466.0050
1.06466.0500
CA1.06466.0050EA
266
CAD
CA1.06466.0050
CA1.06466.0500
Sodium hydroxide monohydrate, Suprapur® for trace metal analysis, Supelco®
Sodium hydroxide monohydrate
Formula:
HNaO·1H₂O MW: 58.012 g/mol |
MDL Number:
MFCD00149229 CAS Number: 12179-02-1 EINECS: 215-185-5 UN: 1823 ADR: 8 II |
Specification Test Results
Solubility | 1100 g/l (20 °C) |
Molar Mass | 58.01 g/mol |
Boiling Point | 1400 °C |
pH | 14 (50 g/l, H2O, 20 °C) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...