Sodium fluoride ≥99.5% (by precipitation titration), EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06449.0250 1.06449.1000 1.06449.9050
CA1.06449.0250EA 194 CAD
CA1.06449.0250 CA1.06449.1000 CA1.06449.9050
Sodium fluoride ≥99.5% (by precipitation titration), EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Sodium fluoride
Formula: NaF
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 996 °C
Density: 2.8 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (precipitative titration) ≥ 99.5 %
In water insoluble matter ≤ 0.01 %
Free alkali (as NaOH) ≤ 0.04 %
Free acid (as hydrofloric acid) ≤ 0.05 %
Chloride (Cl) ≤ 0.01 %
Hexafluorosilicate (SiF₆) ≤ 0.075 %
Sulfate (SO₄) ≤ 0.01 %
Sulfite (SO₃) ≤ 0.005 %
Heavy metals (as Pb) ≤ 0.001 %
Ca (Calcium) ≤ 0.004 %
Cu (Copper) ≤ 0.0002 %
Fe (Iron) ≤ 0.002 %
K (Potassium) ≤ 0.02 %
Loss on drying (150 °C, 4 h) ≤ 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR