tri-Sodium citrate dihydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

1.06448.0500 1.06448.1000 1.06448.5000
CA1.06448.0500EA 157 CAD
CA1.06448.0500 CA1.06448.1000 CA1.06448.5000
tri-Sodium citrate dihydrate, EMSURE® for analysis, Supelco®
tri-Sodium citrate dihydrate

In North America 1.06448.0500 replaces but has a far superior specification to SX0445-1
In North America 1.06448.1000 replaces but has a far superior specification and different pack size to SX0445-3
In North America 1.06448.5000 replaces but has a far superior specification and different pack size to SX0445-3 and SX0445-5
In North America 1.06448.9025 replaces but has a far superior specification and different pack size to SX0445-20

Formula: Na₃C₆H₅O₇·2H₂O
Melting Pt: 150 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 425 g/l (25 °C)
Melting Point 150 °C (anhydrous substance)
Molar Mass 294.10 g/mol
Bulk Density 900 - 1100 kg/m3
Density 1.76 g/cm3
pH 7.5 - 9.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR