Sodium carbonate, anhydrous ≥99.9% (by acidimetry on anhydrous substance), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.06393.1000 1.06393.9050
CA1.06393.1000EA 349 CAD
CA1.06393.1000 CA1.06393.9050
Sodium carbonate, anhydrous ≥99.9% (by acidimetry on anhydrous substance), EMSURE® for analysis, Supelco®
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 851 °C (lit.)
Density: 2.53 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 220 g/l (20 °C)
Melting Point 854 °C
Molar Mass 105.99 g/mol
Bulk Density 1100 kg/m3
Boiling Point 1600 °C (decomposition)
Density 2.53 g/cm3 (20 °C)
pH 11.5 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR