Sodium carbonate decahydrate 99-102% (by acidimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Soda

1.06391.1000 1.06391.5000 1.06391.9025
CA1.06391.1000EA 159 CAD
CA1.06391.1000 CA1.06391.5000 CA1.06391.9025
Sodium carbonate decahydrate 99-102% (by acidimetry), EMSURE® for analysis, Supelco®
Sodium carbonate decahydrate
Formula: Na₂CO₃·10H₂O
Melting Pt: 33 °C (–H₂O of crystall.)
Density: 1.44 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8

Order Now

Specification Test Results

Solubility 210 g/l (20 °C)
Melting Point 33 °C Elimination of water of crystallisation
Molar Mass 286.14 g/mol
Bulk Density 700 - 900 kg/m3
Density 1.44 g/cm3 (20 °C)
pH 11 - 12 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR