Sodium hydrogen carbonate 99.0-101.0% (by acidimetry) [99.7-100.3% dry basis (acidimetric)], EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Synonyms:
Sodium bicarbonate
1.06329.0500
1.06329.1000
1.06329.5000
1.06329.9025
1.06329.9050
CA1.06329.0500EA
102
CAD
CA1.06329.0500
CA1.06329.1000
CA1.06329.5000
CA1.06329.9025
CA1.06329.9050
Sodium hydrogen carbonate 99.0-101.0% (by acidimetry) [99.7-100.3% dry basis (acidimetric)], EMSURE® for analysis, Supelco®
Sodium hydrogen carbonate
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 (dec.) Density: 2.22 g/cm3 (20 °C), 2.16 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
|
Specification Test Results
Solubility | 96 g/l (20 °C) |
Melting Point | 270 °C (decomposition) |
Molar Mass | 84.01 g/mol |
Bulk Density | 1000 kg/m3 |
Vapor Pressure | 8.3 hPa (30 °C) |
Density | 2.22 g/cm3 (20 °C) |
pH | 8.6 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...