Alizarin red S (sodium salt) for analysis, indicator (C.I. 58005), Supelco®

Supplier: MilliporeSigma
Warning

1.06278.0025 1.06278.0100 1.06278.0025 1.06278.0100
EM1.06278.0025CS 9187.7 CAD
EM1.06278.0025 EM1.06278.0100 CA1.06278.0025 CA1.06278.0100
Alizarin red S (sodium salt) for analysis, indicator (C.I. 58005), Supelco®
Alizarin Red S (sodium salt)
Formula: C₁₄H₇NaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
EINECS: 204-981-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 342.26 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR