Sodium acetate, anhydrous 99,0 Perchloric acid titration, EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma

Synonyms: Acetic acid sodium salt

1.06268.1000 1.06268.2500
CA1.06268.1000EA 241 CAD
CA1.06268.1000 CA1.06268.2500
Sodium acetate, anhydrous 99,0 Perchloric acid titration, EMSURE® analysis_for, Supelco®
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4 (dec.)
Melting Pt: 324 (dec.)
Density: 1.52 g/cm3 (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Solubility 365 g/l (20 °C)
Melting Point 324 °C (decomposition)
Molar Mass 82.04 g/mol
Boiling Point 400 °C (decomposition)
Vapor Pressure (20 °C)
Flash Point 250 °C
Density 1.52 g/cm3 (20 °C)
pH 7.5 - 9.0 (50 g/l, H2O, 20 °C)
Ignition Point 607 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR