Sodium acetate trihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% dry basis (perchloric acid titration)], EMSURE® for analysis indifferent to potassium permanganate, Supelco®

Supplier: MilliporeSigma

Synonyms: Acetic acid sodium salt trihydrate

1.06267.0500 1.06267.1000 1.06267.5000 1.06267.9050
CA1.06267.0500EA 137 CAD
CA1.06267.0500 CA1.06267.1000 CA1.06267.5000 CA1.06267.9050
Sodium acetate trihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% dry basis (perchloric acid titration)], EMSURE® for analysis indifferent to potassium permanganate, Supelco®
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120 - 123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.42 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 613 g/l (20 °C)
Melting Point 58 °C
Molar Mass 136.08 g/mol
Bulk Density 900 kg/m3
Boiling Point 400 °C (anhydrous substance),(decomposition)
Vapor Pressure (20 °C)
Flash Point 250 °C (anhydrous substance)
Density 1.42 g/cm3 (20 °C)
pH 8 - 9.5 (10 g/l, H2O, 20 °C)
Ignition Point 607 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR