Methylthymol blue sodium salt indicator for metal titration, Supelco®
Supplier: MilliporeSigma
Synonyms:
Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate, Methylthymol Blue tetrasodium salt
1.06084.0001
1.06084.0005
1.06084.0001
1.06084.0005
EM1.06084.0001CS
25565.98
CAD
EM1.06084.0001
EM1.06084.0005
CA1.06084.0001
CA1.06084.0005
Methylthymol blue sodium salt indicator for metal titration, Supelco®
Methylthymol Blue sodium salt
Formula:
C₃₇H₄₀N₂Na₄O₁₃S MW: 844.76 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00148905 CAS Number: 1945-77-3 EINECS: 217-743-3 |
Specification Test Results
Solubility | 460 g/l (20 °C) |
Melting Point | 844.76 g/mol |
Molar Mass | 570 kg/m3 |
pH | 6.2 (10 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...