Methyl red indicator (C.I. 13020), Supelco®

Supplier: MilliporeSigma

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

1.06076.0100 1.06076.0100 1.06076.0025
EM1.06076.0100CS 1959.2 CAD
EM1.06076.0100 CA1.06076.0100 CA1.06078.0025
Methyl red indicator (C.I. 13020), Supelco®
Methyl red
Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Melting Pt: 178…182 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 178 - 182 °C
Molar Mass 269.31 g/mol
Bulk Density 300 - 500 kg/m3
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR