Manganese(II) nitrate tetrahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.05940.0500 1.05940.1000
CA1.05940.0500EA 242 CAD
CA1.05940.0500 CA1.05940.1000
Manganese(II) nitrate tetrahydrate, EMSURE® for analysis, Supelco®
Manganese (II) nitrate tetrahydrate
Formula: Mn(NO₃)₂·4H₂O
Melting Pt: 37 °C
Density: 2.115 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149789
CAS Number: 20694-39-7
EINECS: 233-828-8
UN: 2724
ADR: 5.1,III

Order Now

Specification Test Results

Solubility 3800 g/l (20 °C)
Melting Point 37 °C
Molar Mass 251.01 g/mol
Bulk Density 750 - 950 kg/m3
Density 2.13 g/cm3 (20 °C)
pH 3 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR