Manganese(II) chloride tetrahydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

1.05927.0100 1.05927.1000
CA1.05927.0100EA 84 CAD
CA1.05927.0100 CA1.05927.1000
Manganese(II) chloride tetrahydrate, EMSURE® ACS for analysis, Supelco®
Manganese (II) chloride tetrahydrate
Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3288
ADR: 6.1 III
Merck Index: 13,05751

Order Now

Specification Test Results

Assay (complexometric) 99.0 - 101.0 %
Insoluble matter ≤ 0.005 %
pH-value (5 %; water; 25 °C) 3.5 - 6.0
Sulfate (SO₄) ≤ 0.002 %
Heavy metals (as Pb) ≤ 0.0005 %
Ca (Calcium) ≤ 0.001 %
Cu (Copper) ≤ 0.0005 %
Fe (Iron) ≤ 0.0005 %
K (Potassium) ≤ 0.001 %
Mg (Magnesium) ≤ 0.005 %
Na (Sodium) ≤ 0.005 %
Pb (Lead) ≤ 0.0005 %
Zn (Zinc) ≤ 0.001 %
Substances not precipitated by ammonium sulfide (as SO₄) ≤ 0.2 %
Substances reducing potassium permanganate (as O) ≤ 0.0005 %
Corresponds to ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR