Magnesium perchlorate hydrate ∼ 83% Mg(ClO₄)₂ ∼ 83% Mg(ClO₄)₂, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Anhydrone hydrate, Magnesium perchlorate hydrat

1.05874.0100 1.05874.0500
CA1.05874.0100EA 446 CAD
CA1.05874.0100 CA1.05874.0500
Magnesium perchlorate hydrate ∼ 83% Mg(ClO₄)₂ ∼ 83% Mg(ClO₄)₂, EMSURE® for analysis, Supelco®
Magnesium perchlorate hydrate
Formula: Cl₂MgO₈·H₂O
Melting Pt: 146 °C
Density: 2.6 g/cm³ (25 °C) (anhydr.)
Storage Temperature: Ambient
CAS Number: 64010-42-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II

Order Now

Specification Test Results

Solubility 500 g/l (25 °C)
Melting Point 251 °C decomposes
Bulk Density 600 - 800 kg/m3
Density 2.60 g/cm3 (25 °C) (anhydrous substance)
pH 8.2 (50 g/l, H2O, 25 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR