Magnesium nitrate hexahydrate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.05855.0050 1.05855.0500
CA1.05855.0050EA 453 CAD
CA1.05855.0050 CA1.05855.0500
Magnesium nitrate hexahydrate, Suprapur® for trace metal analysis, Supelco®
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Boiling Pt: 330
Melting Pt: 89
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 93.3
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
UN: 1474
Merck Index: 13,05697

Order Now

Specification Test Results

Solubility 420 g/l (20 °C)
Melting Point 89 °C (for the hexahydrate)
Molar Mass 256.41 g/mol
Density 1.46 g/cm3 (20 °C)
pH 5.0 - 7.0 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR