di-Lithium tetraborate analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Lithium tetraborate, Dilithium tetraborate

1.05699.0100 1.05699.1000
CA1.05699.0100EA 107 CAD
CA1.05699.0100 CA1.05699.1000
di-Lithium tetraborate analysis_for, Supelco®
di-Lithium tetraborate

EMD # 1.05699.1000 replaces and is identical to EMD # LX0366A-2.

Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524

Order Now

Specification Test Results

Assay (acidimetric) ≥ 98.0 %
Chloride (Cl) ≤ 0.005 %
Phosphate (PO₄) ≤ 0.002 %
Sulfate (SO₄) ≤ 0.005 %
Heavy metals (as Pb) ≤ 0.005 %
Ca (Calcium) ≤ 0.005 %
Fe (Iron) ≤ 0.001 %
K (Potassium) ≤ 0.005 %
Mg (Magnesium) ≤ 0.001 %
Na (Sodium) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR