Lithium fluoride, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.05686.0050
1.05686.0050
1.05686.0500
1.05686.0500
CA1.05686.0500EA
1754.83
CAD
EM1.05686.0500
EM1.05686.0050
CA1.05686.0500
CA1.05686.0050
Lithium fluoride, Suprapur® for trace metal analysis, Supelco®
Lithium fluoride
Formula:
FLi MW: 25.94 g/mol Boiling Pt: 1676 °C (1013 hPa) Melting Pt: 870 °C Density: 2.64 g/cm³ (20 °C) |
MDL Number:
MFCD00011090 CAS Number: 7789-24-4 UN: 3288 ADR: 6.1,III Merck Index: 13,05553 |
Specification Test Results
Solubility | 1.3 g/l (25 °C) |
Melting Point | 870 °C |
Molar Mass | 25.94 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 1676 °C (1013 hPa) |
Density | 2.64 g/cm3 (20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Hazard Class Description | Toxic Materials |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...