Lithium chloride, EMSURE® analysis_for, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.05679.0100
1.05679.0250
CA1.05679.0100EA
318
CAD
CA1.05679.0100
CA1.05679.0250
Lithium chloride, EMSURE® analysis_for, Supelco®
Lithium chloride
Formula:
LiCl MW: 42.39 g/mol Boiling Pt: 1382 Melting Pt: 614 Density: 2.07 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 EINECS: 231-212-3 Merck Index: 13,05550 |
Specification Test Results
Solubility | 832 g/l (20 °C) |
Melting Point | 614 °C |
Molar Mass | 42.39 g/mol |
Bulk Density | 530 kg/m3 |
Boiling Point | 1360 °C (1013 hPa) |
Vapor Pressure | 1.33 hPa (547 °C) |
Density | 2.07 g/cm3 (20 °C) |
pH | 6 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...